Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177310.1 | NP_001170781.1 | 3 | 188 | ||
NM_001177311.1 | NP_001170782.1 | 3 | 215 | |||
NM_017662.4 | NP_060132.3 | 3 | 369 | |||
XM_011518244.1 | XP_011516546.1 | 3 | 369 | |||
XM_011518245.1 | XP_011516547.1 | 3 | 369 | |||
XM_011518246.1 | XP_011516548.1 | 3 | 369 | |||
XM_011518247.1 | XP_011516549.1 | 3 | 369 | |||
XM_011518248.1 | XP_011516550.1 | 3 | 369 | |||
XM_011518249.1 | XP_011516551.1 | 3 | 369 | |||
XM_011518250.1 | XP_011516552.1 | 3 | 369 | |||
XM_011518252.2 | XP_011516554.1 | 3 | 369 | |||
XM_011518255.2 | XP_011516557.1 | 3 | 369 | |||
XM_017014287.1 | XP_016869776.1 | 3 | 369 | |||
XM_017014288.1 | XP_016869777.1 | 3 | 369 | |||
XM_017014289.1 | XP_016869778.1 | 3 | 369 | |||
GenBank mRNA | AF350881.2 | - | 3 | 369 | ||
AF448232.1 | - | 3 | 131 | |||
AK057748.1 | - | 3 | 369 | |||
AY333282.1 | - | 3 | 194 | |||
AY333283.1 | - | 3 | 188 | |||
AY333284.1 | - | 3 | 215 | |||
AY333285.1 | - | 3 | 194 | |||
AY333286.1 | - | 3 | 194 | |||
AY333287.1 | - | 3 | 194 | |||
AY333288.1 | - | 3 | 194 |