Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207076.2 | NP_001194005.1 | 4 | 573 | ||
NM_080834.3 | NP_543024.2 | 4 | 573 | |||
NR_130714.1 | 4 | 573 | ||||
XM_005260295.2 | XP_005260352.1 | 4 | 547 | |||
XM_011528590.2 | XP_011526892.1 | 4 | 533 | |||
XM_011528592.1 | XP_011526894.1 | 3 | 403 | |||
XM_011528598.1 | XP_011526900.1 | 1 | 60 | |||
XM_017027683.1 | XP_016883172.1 | 4 | 751 | |||
GenBank mRNA | AK057454.1 | - | 4 | 547 | ||
BC114634.1 | - | 4 | 531 | |||
BC114945.1 | - | 4 | 531 | 40084218 | ||
HY055403.1 | - | 4 | 447 |