- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: VTI1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318203.1 | NP_001305132.1 | 4 | 669 | ||
NM_001318205.1 | NP_001305134.1 | 4 | 669 | |||
NM_145206.3 | NP_660207.2 | 4 | 669 | |||
XM_005269544.4 | XP_005269601.1 | 4 | 623 | |||
XM_005269545.4 | XP_005269602.1 | 4 | 669 | |||
XM_005269546.3 | XP_005269603.1 | 4 | 669 | |||
XM_005269547.4 | XP_005269604.1 | 4 | 623 | |||
XM_006717637.2 | XP_006717700.1 | 4 | 669 | |||
XM_011539328.2 | XP_011537630.1 | 4 | 669 | |||
XM_011539330.2 | XP_011537632.1 | 4 | 669 | |||
XM_011539331.2 | XP_011537633.1 | 4 | 669 | |||
XM_011539333.2 | XP_011537635.1 | 4 | 669 | |||
XM_017015745.1 | XP_016871234.1 | 4 | 669 | |||
XM_017015746.1 | XP_016871235.1 | 4 | 623 | |||
XM_017015747.1 | XP_016871236.1 | 4 | 669 | |||
XM_017015748.1 | XP_016871237.1 | 4 | 623 | |||
GenBank mRNA | AK303646.1 | - | 4 | 380 | ||
BC017052.1 | - | 4 | 399 | 3885564 | ||
BI010424.1 | - | 5 | 378 | |||
BI909044.1 | - | 4 | 427 |