Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001872.3 | NP_001001872.2 | 2 | 1251 | ||
NM_001320173.1 | NP_001307102.1 | 2 | 1251 | |||
XM_011536471.2 | XP_011534773.1 | 2 | 1251 | |||
XM_017021029.1 | XP_016876518.1 | 2 | 1108 | |||
XM_017021030.1 | XP_016876519.1 | 2 | 1251 | |||
GenBank mRNA | AK292513.1 | - | 2 | 1225 | ||
AK292605.1 | - | 2 | 1168 | |||
AK294599.1 | - | 1 | 462 | |||
BC013707.1 | - | 1 | 548 | |||
BC146861.1 | - | 2 | 1102 | 9056789 | ||
BX161411.1 | - | 2 | 1141 | |||
BX161448.1 | - | 2 | 1195 | |||
BX649130.1 | - | 1 | 747 |