Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033576.1 | NP_001028748.1 | 8 | 1033 | ||
NM_001267052.1 | NP_001253981.1 | 8 | 1030 | |||
NM_001308281.1 | NP_001295210.1 | 7 | 936 | |||
NM_173167.3 | NP_775259.1 | 7 | 936 | |||
XM_017024234.1 | XP_016879723.1 | 8 | 1054 | |||
GenBank mRNA | AF539794.1 | - | 7 | 936 | ||
AL832355.1 | - | 8 | 1027 | |||
AL833281.1 | - | 8 | 1006 | |||
BC101062.2 | - | 7 | 936 | 40008186 | ||
BC101063.2 | - | 7 | 936 | 40008187 | ||
BX647642.1 | - | 8 | 1033 |