Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178002.2 | NP_001171473.1 | 27 | 3641 | ||
NM_138813.3 | NP_620168.1 | 27 | 3732 | |||
NR_047593.2 | 27 | 3876 | ||||
XM_006722654.2 | XP_006722717.1 | 27 | 3762 | |||
XM_006722655.2 | XP_006722718.1 | 27 | 3841 | |||
XM_006722656.2 | XP_006722719.1 | 27 | 3650 | |||
XM_011527707.1 | XP_011526009.1 | 27 | 3735 | |||
XM_011527708.1 | XP_011526010.1 | 27 | 3642 | |||
XM_011527709.1 | XP_011526011.1 | 26 | 3549 | |||
XM_011527710.2 | XP_011526012.1 | 21 | 2971 | |||
XM_011527712.1 | XP_011526014.1 | 12 | 1764 | |||
GenBank mRNA | AK057452.1 | - | 4 | 592 | ||
AK125969.1 | - | 27 | 3876 | |||
AY302538.1 | - | 27 | 3604 | |||
BC035162.2 | - | 27 | 3727 | 5265541 |