Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280799.1 | NP_001267728.1 | 8 | 2583 | ||
NM_144698.4 | NP_653299.4 | 10 | 2853 | |||
GenBank mRNA | AK057853.1 | - | 1 | 743 | ||
AK091120.1 | - | 10 | 2818 | |||
AK294308.1 | - | 8 | 2562 | |||
BC101618.1 | - | 10 | 2800 | 8069124 | ||
BC101620.1 | - | 10 | 2800 | 8069126 |