Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152490.4 | NP_689703.1 | 8 | 1208 | ||
XM_006711749.3 | XP_006711812.1 | 8 | 1150 | |||
XM_017000394.1 | XP_016855883.1 | 9 | 1331 | |||
XM_017000395.1 | XP_016855884.1 | 9 | 1284 | |||
GenBank mRNA | AB209046.1 | - | 7 | 762 | ||
AK311626.1 | - | 3 | 480 | |||
BC029564.1 | - | 8 | 1208 | 4826983 |