Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145454.2 | NP_001138926.1 | 5 | 1667 | ||
GenBank mRNA | AK056768.1 | - | 2 | 1207 | ||
BC028398.1 | - | 1 | 1101 | |||
BC114443.1 | - | 2 | 1155 | 40083863 | ||
BC114502.1 | - | 2 | 1155 | 40083868 |