Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYHIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152501.4 | NP_689714.2 | 7 | 1504 | ||
NM_198928.4 | NP_945146.1 | 7 | 1477 | |||
NM_198929.4 | NP_945147.1 | 7 | 1504 | |||
NM_198930.3 | NP_945148.1 | 7 | 1477 | |||
XM_011509242.2 | XP_011507544.1 | 7 | 1504 | |||
XM_011509243.2 | XP_011507545.1 | 7 | 1504 | |||
XM_017000463.1 | XP_016855952.1 | 7 | 1504 | |||
GenBank mRNA | AY185344.1 | - | 7 | 1491 | ||
AY185345.1 | - | 7 | 1464 | |||
AY185346.1 | - | 7 | 1491 | |||
AY185347.1 | - | 7 | 1464 | |||
BC090944.1 | - | 7 | 1486 | 5212268 | ||
BC139741.1 | - | 7 | 1431 | 40033401 |