Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308069.1 | NP_001294998.1 | 3 | 357 | ||
NM_152531.4 | NP_689744.3 | 3 | 820 | |||
XM_005269286.4 | XP_005269343.1 | 3 | 3205 | |||
XM_011512448.2 | XP_011510750.1 | 4 | 632 | |||
XM_017005749.1 | XP_016861238.1 | 3 | 813 | |||
XM_017005750.1 | XP_016861239.1 | 2 | 3593 | |||
GenBank mRNA | AK057046.1 | - | 4 | 565 | ||
AK075551.1 | - | 3 | 788 | |||
AK301133.1 | - | 3 | 293 | |||
BC019036.2 | - | 3 | 729 | 4591786 | ||
BC039067.1 | - | 3 | 802 | 5185557 | ||
BG721419.1 | - | 3 | 362 |