Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 14 | 2073 | ||
NM_153223.3 | NP_694955.2 | 15 | 2154 | |||
XM_005271901.4 | XP_005271958.1 | 14 | 1935 | |||
XM_011543185.2 | XP_011541487.1 | 14 | 2488 | |||
XM_011543186.2 | XP_011541488.1 | 11 | 1767 | |||
XM_017009085.1 | XP_016864574.1 | 11 | 1632 | |||
GenBank mRNA | AK093409.1 | - | 7 | 840 | ||
AK095646.1 | - | 14 | 2075 | |||
AL833929.1 | - | 3 | 261 | |||
BC040527.1 | - | 14 | 2737 | |||
BC144558.1 | - | 17 | 2281 | |||
BX648687.1 | - | 5 | 547 |