Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC23L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317949.1 | NP_001304878.1 | 3 | 267 | ||
NM_144725.3 | NP_653326.3 | 3 | 332 | |||
XM_011513984.2 | XP_011512286.1 | 3 | 269 | |||
XM_011513985.2 | XP_011512287.1 | 3 | 269 | |||
XM_011513987.2 | XP_011512289.1 | 3 | 269 | |||
XM_017009119.1 | XP_016864608.1 | 3 | 513 | |||
XM_017009120.1 | XP_016864609.1 | 3 | 332 | |||
XM_017009121.1 | XP_016864610.1 | 3 | 377 | |||
XM_017009122.1 | XP_016864611.1 | 3 | 448 | |||
XM_017009123.1 | XP_016864612.1 | 3 | 269 | |||
GenBank mRNA | AK058168.1 | - | 3 | 332 | ||
AY498865.1 | - | 2 | 229 | |||
BC057775.1 | - | 3 | 304 | 5295348 | ||
JN799997.1 | - | 3 | 267 |