Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144727.2 | NP_653328.1 | 3 | 535 | ||
NR_131779.1 | 3 | 887 | ||||
XM_005249952.3 | XP_005250009.1 | 3 | 535 | |||
XM_011515836.1 | XP_011514138.1 | 3 | 535 | |||
XM_011515837.2 | XP_011514139.1 | 3 | 914 | |||
XM_017011773.1 | XP_016867262.1 | 3 | 692 | |||
GenBank mRNA | AF445455.1 | - | 3 | 378 | ||
BC043605.1 | - | 3 | 887 | |||
BC100878.2 | - | 3 | 373 | 40003204 | ||
BC100880.2 | - | 3 | 373 | 40003208 | ||
BC100881.2 | - | 3 | 373 | 40003209 |