Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287049.1 | NP_001273978.1 | 4 | 1474 | ||
NM_001287050.1 | NP_001273979.1 | 3 | 1513 | |||
NM_153707.3 | NP_714918.2 | 4 | 1716 | |||
XM_011517744.1 | XP_011516046.1 | 5 | 1796 | |||
XM_011517745.1 | XP_011516047.1 | 6 | 1731 | |||
XM_011517746.2 | XP_011516048.1 | 6 | 1652 | |||
XM_011517747.2 | XP_011516049.1 | 3 | 1312 | |||
XM_017014314.1 | XP_016869803.1 | 6 | 2042 | |||
XM_017014315.1 | XP_016869804.1 | 4 | 1429 | |||
GenBank mRNA | AA992255.1 | - | 1 | 218 | ||
AI808568.1 | - | 1 | 464 | |||
AK057474.1 | - | 3 | 1508 | |||
AK292245.1 | - | 4 | 1667 | |||
AW079914.1 | - | 1 | 349 | |||
BC033489.1 | - | 4 | 1644 | 5170510 |