Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
siRNA ID
HSS136159
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
18
3382
NM_152586.3
NP_689799.3
18
3480
NR_135249.1
19
3867
NR_135250.1
14
2862
XM_005269582.2
XP_005269639.1
17
6558
XM_011539368.2
XP_011537670.1
18
6624
XM_017015759.1
XP_016871248.1
20
6053
XM_017015760.1
XP_016871249.1
20
4955
XM_017015761.1
XP_016871250.1
20
5793
XM_017015762.1
XP_016871251.1
20
4963
XM_017015763.1
XP_016871252.1
20
4257
XM_017015764.1
XP_016871253.1
20
4320
XM_017015765.1
XP_016871254.1
21
4573
XM_017015766.1
XP_016871255.1
20
6179
XM_017015767.1
XP_016871256.1
20
5838
XM_017015768.1
XP_016871257.1
20
4009
XM_017015769.1
XP_016871258.1
20
3617
XM_017015770.1
XP_016871259.1
20
4278
XM_017015771.1
XP_016871260.1
19
6795
XM_017015772.1
XP_016871261.1
19
6801
XM_017015773.1
XP_016871262.1
18
6735
XM_017015774.1
XP_016871263.1
19
5721
XM_017015775.1
XP_016871264.1
20
5599
XM_017015776.1
XP_016871265.1
19
6104
XM_017015777.1
XP_016871266.1
19
5763
XM_017015778.1
XP_016871267.1
18
6681
XM_017015779.1
XP_016871268.1
19
6801
XM_017015780.1
XP_016871269.1
19
6801
XM_017015781.1
XP_016871270.1
18
6630
XM_017015782.1
XP_016871271.1
19
4381
XM_017015783.1
XP_016871272.1
19
5428
XM_017015784.1
XP_016871273.1
18
3374
XM_017015785.1
XP_016871274.1
18
6681
XM_017015786.1
XP_016871275.1
18
3871
GenBank mRNA
AK094637.1
-
1
574
AK126497.1
-
6
1835
AK127213.1
-
5
1415
AK127267.1
-
14
2862
AK303633.1
-
3
1073
AL832102.1
-
1
520
BC026037.1
-
1
328
BC064967.1
-
1
516
BC130633.1
-
3
1064
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0