Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145913.3 | NP_666018.3 | 12 | 1765 | ||
GenBank mRNA | AF536216.1 | - | 12 | 1738 | ||
AK075263.1 | - | 12 | 1423 | |||
AK313788.1 | - | 12 | 1734 | |||
AY081220.1 | - | 12 | 1739 | |||
BC110492.2 | - | 12 | 1400 | 40007567 |