Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258205.1 | NP_001245134.1 | 13 | 1925 | ||
NM_145068.3 | NP_659505.1 | 13 | 1925 | |||
GenBank mRNA | AF514998.1 | - | 13 | 1667 | ||
AJ487035.2 | - | 13 | 1925 | |||
AK127726.1 | - | 7 | 877 | |||
AY118267.1 | - | 13 | 1735 | |||
AY118268.1 | - | 13 | 1735 | |||
BC104866.1 | - | 13 | 1697 | 8143869 | ||
BC104868.1 | - | 13 | 1697 | 8143871 | ||
BC143294.1 | - | 14 | 1866 | 9051797 | ||
BC143299.1 | - | 14 | 1866 | 9051803 | ||
BC143330.1 | - | 13 | 1697 | 9051835 | ||
BC143333.1 | - | 14 | 1786 | |||
BC143334.1 | - | 13 | 1779 | |||
EU295940.1 | - | 12 | 1603 |