Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199085.1 | NP_001186014.1 | 4 | 1211 | ||
NM_001199089.1 | NP_001186018.1 | 4 | 924 | |||
NM_001199091.1 | NP_001186020.1 | 4 | 1052 | |||
NM_173533.3 | NP_775804.2 | 4 | 1211 | |||
XM_005244934.1 | XP_005244991.1 | 4 | 996 | |||
XM_005244935.4 | XP_005244992.1 | 4 | 901 | |||
XM_017000473.1 | XP_016855962.1 | 4 | 917 | |||
GenBank mRNA | AK126889.1 | - | 4 | 1051 | ||
AK127370.1 | - | 4 | 943 | |||
BC065707.1 | - | 1 | 20 | |||
BC089387.1 | - | 2 | 113 | |||
BC130532.1 | - | 4 | 888 | 40146563 | ||
BC144058.1 | - | 4 | 943 | 9052578 |