Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139056.3 | NP_620687.2 | 18 | 2838 | ||
NR_136935.1 | 17 | 2707 | ||||
GenBank mRNA | AB095949.1 | - | 15 | 2093 | ||
AJ315734.1 | - | 18 | 2700 |