Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139057.3 | NP_620688.2 | 10 | 1557 | ||
XM_005254872.3 | XP_005254929.1 | 10 | 2473 | |||
XM_011521312.2 | XP_011519614.1 | 10 | 2473 | |||
XM_017021973.1 | XP_016877462.1 | 11 | 2606 | |||
XM_017021974.1 | XP_016877463.1 | 11 | 2608 | |||
XM_017021975.1 | XP_016877464.1 | 11 | 2606 | |||
XM_017021976.1 | XP_016877465.1 | 10 | 1318 | |||
XM_017021977.1 | XP_016877466.1 | 11 | 2605 | |||
XM_017021978.1 | XP_016877467.1 | 4 | 3864 | |||
XM_017021979.1 | XP_016877468.1 | 3 | 314 | |||
XM_017021980.1 | XP_016877469.1 | 3 | 524 | |||
XM_017021981.1 | XP_016877470.1 | 11 | 2603 | |||
XM_017021983.1 | XP_016877472.1 | 1 | 111 | |||
XM_017021984.1 | XP_016877473.1 | 9 | 1132 | |||
GenBank mRNA | AJ315735.1 | - | 10 | 1474 | ||
AK131344.1 | - | 9 | 1103 |