Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276700.1 | NP_001263629.1 | 2 | 90 | ||
NM_138329.2 | NP_612202.2 | 2 | 90 | |||
XM_011519931.1 | XP_011518233.1 | 2 | 295 | |||
XM_017017252.1 | XP_016872741.1 | 2 | 295 | |||
GenBank mRNA | AF479748.1 | - | 2 | 90 | ||
AK292668.1 | - | 2 | 295 | |||
AY154461.1 | - | 2 | 90 |