Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM129C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098524.1 | NP_001091994.1 | 12, 13 | 1677 | ||
NM_001321826.1 | NP_001308755.1 | 11, 12 | 1549 | |||
NM_001321827.1 | NP_001308756.1 | 11, 12 | 1549 | |||
NM_001321828.1 | NP_001308757.1 | 10, 11 | 1387 | |||
NM_173544.4 | NP_775815.2 | 12, 13 | 1677 | |||
XM_005259813.3 | XP_005259870.1 | 11, 12 | 1524 | |||
XM_011527781.2 | XP_011526083.1 | 11, 12 | 1524 | |||
XM_011527786.2 | XP_011526088.1 | 11, 12 | 1524 | |||
XM_011527787.2 | XP_011526089.1 | 11, 12 | 1524 | |||
XM_011527789.1 | XP_011526091.1 | 11, 12 | 1524 | |||
XM_017026453.1 | XP_016881942.1 | 11, 12 | 1524 | |||
XM_017026455.1 | XP_016881944.1 | 10, 11 | 1396 | |||
XM_017026456.1 | XP_016881945.1 | 10, 11 | 1451 | |||
XM_017026457.1 | XP_016881946.1 | 8, 9 | 1101 | |||
GenBank mRNA | AK074069.1 | - | 11, 12 | 1499 | ||
AK097121.1 | - | 11, 12 | 1524 | |||
AK298062.1 | - | 10, 11 | 1378 | |||
AK300046.1 | - | 11, 12 | 1531 | |||
AK301159.1 | - | 11, 12 | 1427 | |||
AY254197.1 | - | 12, 13 | 1677 | |||
AY254198.1 | - | 12, 13 | 1677 | |||
AY254199.1 | - | 12, 13 | 1677 |