Search Thermo Fisher Scientific
Gene Symbol: GKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182536.2 | NP_872342.2 | 3, 4 | 312 | ||
GenBank mRNA | AF494509.2 | - | 3, 4 | 312 | ||
AJ966788.1 | - | 3, 4 | 233 | |||
AY358664.1 | - | 3, 4 | 287 | |||
AY943908.1 | - | 3, 4 | 235 |