Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128210.1 | NP_001121682.1 | 6 | 1228 | ||
NM_181784.2 | NP_861449.2 | 6 | 1581 | |||
XM_005264201.1 | XP_005264258.1 | 5 | 996 | |||
XM_017003576.1 | XP_016859065.1 | 4 | 1147 | |||
GenBank mRNA | AK025550.1 | - | 2 | 585 | ||
AK056479.1 | - | 6 | 1145 | |||
AK299079.1 | - | 6 | 1081 | |||
AK307714.1 | - | 4 | 727 | |||
AY299090.1 | - | 6 | 1242 | |||
BC029300.1 | - | 6 | 1158 | |||
BC111495.1 | - | 6 | 1569 | |||
BC130292.1 | - | 6 | 1178 | 40146323 | ||
BC136334.1 | - | 6 | 1178 | 9020321 |