Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282506.1 | NP_001269435.1 | 4 | 2227 | ||
NM_001291833.1 | NP_001278762.1 | 5 | 1702 | |||
NM_020790.1 | NP_065841.1 | 5 | 1762 | |||
NM_152673.3 | NP_689886.3 | 5 | 1714 | |||
GenBank mRNA | AB098731.1 | - | 4 | 1965 | ||
AK027314.1 | - | 6 | 1465 | |||
AK075138.1 | - | 5 | 1423 | |||
AK298316.1 | - | 5 | 1887 | |||
AY358449.1 | - | 5 | 1646 | |||
BC029267.1 | - | 5 | 1673 | 3851952 | ||
BC044243.1 | - | 5 | 1601 | 5754328 |