Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291661.1 | NP_001278590.1 | 70 | 11548 | ||
XM_011533466.1 | XP_011531768.1 | 70 | 11533 | |||
XM_011533467.1 | XP_011531769.1 | 70 | 11500 | |||
XM_011533468.1 | XP_011531770.1 | 70 | 11470 | |||
XM_011533469.1 | XP_011531771.1 | 69 | 11392 | |||
XM_017005860.1 | XP_016861349.1 | 70 | 11533 | |||
XM_017005861.1 | XP_016861350.1 | 69 | 12216 | |||
XM_017005862.1 | XP_016861351.1 | 49 | 8108 | |||
GenBank mRNA | AK097746.1 | - | 10 | 1696 | ||
AK128592.1 | - | 18 | 2974 |