Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291661.1 | NP_001278590.1 | 55 | 8923 | ||
XM_011533466.1 | XP_011531768.1 | 55 | 8908 | |||
XM_011533467.1 | XP_011531769.1 | 55 | 8875 | |||
XM_011533468.1 | XP_011531770.1 | 55 | 8845 | |||
XM_011533469.1 | XP_011531771.1 | 55 | 8908 | |||
XM_011533471.2 | XP_011531773.1 | 55 | 8908 | |||
XM_017005860.1 | XP_016861349.1 | 55 | 8908 | |||
XM_017005861.1 | XP_016861350.1 | 54 | 9591 | |||
XM_017005862.1 | XP_016861351.1 | 34 | 5483 | |||
GenBank mRNA | AK128592.1 | - | 3 | 349 |