Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297697.1 | NP_001284626.1 | 11, 12 | 1036 | ||
NM_176816.4 | NP_789786.2 | 10, 11 | 1197 | |||
XM_005248458.4 | XP_005248515.1 | 10, 11 | 1434 | |||
XM_005248459.4 | XP_005248516.1 | 10, 11 | 1236 | |||
XM_005248461.3 | XP_005248518.1 | 9, 10 | 923 | |||
XM_005248463.3 | XP_005248520.1 | 8, 9 | 791 | |||
XM_006714569.3 | XP_006714632.1 | 10, 11 | 1224 | |||
XM_006714570.3 | XP_006714633.1 | 10, 11 | 1008 | |||
XM_011543255.2 | XP_011541557.1 | 10, 11 | 1214 | |||
XM_011543256.2 | XP_011541558.1 | 10, 11 | 1215 | |||
XM_011543257.2 | XP_011541559.1 | 9, 10 | 1131 | |||
XM_011543258.2 | XP_011541560.1 | 10, 11 | 1239 | |||
XM_011543259.2 | XP_011541561.1 | 10, 11 | 1239 | |||
XM_017009206.1 | XP_016864695.1 | 10, 11 | 1429 | |||
XM_017009207.1 | XP_016864696.1 | 10, 11 | 1224 | |||
XM_017009208.1 | XP_016864697.1 | 8, 9 | 1063 | |||
XM_017009210.1 | XP_016864699.1 | 7, 8 | 984 | |||
XM_017009211.1 | XP_016864700.1 | 4, 5 | 466 | |||
GenBank mRNA | AB024691.1 | - | 10, 11 | 1197 | ||
AB024692.1 | - | 9, 10 | 900 | |||
AK299871.1 | - | 11, 12 | 1036 |