Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304428.1 | NP_001291357.1 | 3 | 922 | ||
NM_001304429.1 | NP_001291358.1 | 3 | 915 | |||
NM_001304431.1 | NP_001291360.1 | 3 | 823 | |||
NM_152687.3 | NP_689900.1 | 3 | 760 | |||
GenBank mRNA | AK090960.1 | - | 3 | 694 | ||
BC063534.1 | - | 1 | 452 | 4430075 |