Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152696.3 | NP_689909.2 | 13 | 2789 | ||
NM_181358.2 | NP_852003.1 | 11 | 1509 | |||
NM_198268.2 | NP_938009.1 | 13 | 2789 | |||
NM_198269.2 | NP_938010.1 | 12 | 1700 | |||
XM_005270609.3 | XP_005270666.1 | 13 | 2966 | |||
XM_005270610.4 | XP_005270667.1 | 13 | 2963 | |||
XM_005270611.4 | XP_005270668.1 | 13 | 2966 | |||
XM_005270612.4 | XP_005270669.1 | 13 | 2928 | |||
XM_005270613.4 | XP_005270670.1 | 15 | 2879 | |||
XM_006710443.3 | XP_006710506.1 | 13 | 2864 | |||
XM_011540977.2 | XP_011539279.1 | 12 | 2846 | |||
XM_017000605.1 | XP_016856094.1 | 13 | 2925 | |||
GenBank mRNA | AB014530.1 | - | 4 | 469 | ||
AB089957.1 | - | 13 | 2753 | |||
AK122662.1 | - | 13 | 2787 | |||
AK302726.1 | - | 13 | 2973 | |||
AL833829.1 | - | 12 | 1678 | |||
BC028408.1 | - | 12 | 1705 | 4821915 | ||
BC033012.2 | - | 13 | 2789 | 4830408 | ||
BC036057.1 | - | 11 | 1509 | 5272816 |