Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LVRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173800.4 | NP_776161.3 | 20 | 3033 | ||
GenBank mRNA | AK027809.1 | - | 3 | 285 | ||
AK075131.1 | - | 15 | 1568 | |||
AK291655.1 | - | 20 | 3047 | |||
AY560010.1 | - | 20 | 2921 | |||
BC036440.1 | - | 16 | 1953 | |||
BC045809.1 | - | 15 | 1671 | |||
BC060869.1 | - | 21 | 2960 | 30346049 | ||
BC068560.1 | - | 20 | 2920 | 30341970 | ||
BC070028.1 | - | 16 | 2071 | |||
BC094716.1 | - | 16 | 2071 | |||
BC109022.1 | - | 20 | 2897 | 40008501 | ||
BC109023.1 | - | 20 | 2897 | 40008502 |