Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005210.2 | NP_001005210.1 | 1 | 549 | ||
GenBank mRNA | AK127591.1 | - | 1 | 549 | ||
BC136737.1 | - | 1 | 507 | 9020727 | ||
BC136739.1 | - | 1 | 507 | 9020729 | ||
BC150572.1 | - | 1 | 391 |