Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153367.3 | NP_699198.2 | 1 | 397 | ||
GenBank mRNA | AK075279.1 | - | 1 | 249 | ||
AK308876.1 | - | 1 | 186 | |||
BC028617.1 | - | 1 | 399 | 4821347 | ||
BC039249.1 | - | 1 | 392 | |||
BC053564.1 | - | 1 | 296 | 6193781 |