Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENKUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270383.1 | NP_001257312.1 | 5 | 781 | ||
NM_145010.3 | NP_659447.1 | 5 | 868 | |||
NR_072992.1 | 5 | 868 | ||||
XM_006717389.1 | XP_006717452.1 | 5 | 864 | |||
XM_006717390.1 | XP_006717453.1 | 4 | 687 | |||
XM_011519348.1 | XP_011517650.1 | 5 | 864 | |||
XM_017015830.1 | XP_016871319.1 | 4 | 687 | |||
GenBank mRNA | AK095021.1 | - | 5 | 813 | ||
AK292495.1 | - | 5 | 865 | |||
AY454125.1 | - | 5 | 855 | |||
BC026165.2 | - | 5 | 870 | 4837530 | ||
CR936720.1 | - | 5 | 781 |