Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244889.1 | NP_001231818.1 | 12 | 2466 | ||
NM_170744.4 | NP_734465.2 | 13 | 2499 | |||
XM_011539453.1 | XP_011537755.1 | 14 | 2048 | |||
XM_017015834.1 | XP_016871323.1 | 14 | 2058 | |||
XM_017015835.1 | XP_016871324.1 | 13 | 2851 | |||
GenBank mRNA | AB096256.1 | - | 13 | 2454 | ||
AK022859.1 | - | 4 | 514 | |||
AK094595.1 | - | 6 | 969 | |||
AK128132.1 | - | 13 | 2499 | |||
AY126437.1 | - | 12 | 2074 | |||
AY358351.1 | - | 13 | 2445 |