Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145016.3 | NP_659453.3 | 5 | 459 | ||
XM_011544809.2 | XP_011543111.1 | 5 | 644 | |||
XM_017017337.1 | XP_016872826.1 | 6 | 594 | |||
XM_017017338.1 | XP_016872827.1 | 5 | 1041 | |||
GenBank mRNA | AB207211.1 | - | 4 | 328 | ||
AF426250.1 | - | 5 | 719 | |||
BC016789.1 | - | 6 | 603 | 4091916 | ||
BC021682.1 | - | 5 | 446 | 4079236 |