Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152727.5 | NP_689940.3 | 2 | 390 | ||
GenBank mRNA | AF492484.1 | - | 2 | 341 | ||
AK292269.1 | - | 1 | 299 | |||
AK315394.1 | - | 2 | 293 | |||
BC053658.1 | - | 3 | 464 | 5750192 | ||
BG830959.1 | - | 2 | 336 | |||
BG831872.1 | - | 2 | 338 | |||
CR749617.1 | - | 1 | 51 | |||
DB504175.1 | - | 2 | 391 |