Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304376.2 | NP_001291305.1 | 9 | 945 | ||
NM_001318481.1 | NP_001305410.1 | 9 | 945 | |||
NM_153837.3 | NP_722579.1 | 9 | 1417 | |||
XM_011522949.2 | XP_011521251.1 | 9 | 961 | |||
XM_011522950.2 | XP_011521252.1 | 9 | 987 | |||
GenBank mRNA | AY140956.1 | - | 8 | 842 | ||
BC032401.1 | - | 9 | 1365 | 5210813 |
Set Membership: |
![]() |