Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286834.1 | NP_001273763.1 | 3 | 379 | ||
NM_001286837.1 | NP_001273766.1 | 3 | 617 | |||
NM_002936.4 | NP_002927.2 | 3 | 379 | |||
GenBank mRNA | AF039652.1 | - | 3 | 349 | ||
AF048994.1 | - | 3 | 295 | |||
AF048995.1 | - | 3 | 329 | |||
AK075490.1 | - | 3 | 328 | |||
AK096913.1 | - | 3 | 535 | |||
BC002973.1 | - | 3 | 338 | 3537074 | ||
BT019670.1 | - | 3 | 268 | |||
CR541820.1 | - | 3 | 268 | |||
DA532761.1 | - | 3 | 379 |