Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCO4 SIRNA Assays ›
Gene Symbol:
TMCO4
Gene Name:
transmembrane and coiled-coil domains 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 19682213 - 19800385 on Build GRCh38
UniGene ID:
Hs.656313
Species:
Human
siRNA ID
HSS137892
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCO4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181719.4
NP_859070.3
8
775
XM_005245822.4
XP_005245879.1
8
791
XM_005245824.4
XP_005245881.1
8
855
XM_005245825.4
XP_005245882.1
7
656
XM_005245827.3
XP_005245884.1
8
791
XM_006710551.3
XP_006710614.1
8
880
XM_011541179.2
XP_011539481.1
11
1250
XM_011541180.2
XP_011539482.1
11
1405
XM_011541181.1
XP_011539483.1
10
1079
XM_011541185.2
XP_011539487.1
8
798
XM_011541186.2
XP_011539488.1
8
895
XM_017000913.1
XP_016856402.1
8
799
XM_017000914.1
XP_016856403.1
10
1299
XM_017000915.1
XP_016856404.1
10
1021
XM_017000916.1
XP_016856405.1
8
933
XM_017000917.1
XP_016856406.1
9
1148
XM_017000918.1
XP_016856407.1
10
1345
XM_017000919.1
XP_016856408.1
9
1072
XM_017000920.1
XP_016856409.1
8
1290
XM_017000921.1
XP_016856410.1
9
1097
XM_017000922.1
XP_016856411.1
7
756
GenBank mRNA
AK309310.1
-
8
791
AK309900.1
-
6
590
AL049846.1
-
5
541
AL049847.1
-
5
539
AX775873.1
-
7
701
AX775875.1
-
7
804
AX775877.1
-
7
804
BC053600.1
-
6
634
5744211
BX640987.1
-
8
776
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0