Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114759.2 | NP_001108231.1 | 5 | 1202 | ||
NM_001307925.1 | NP_001294854.1 | 5 | 1199 | |||
NM_173574.3 | NP_775845.2 | 5 | 1199 | |||
XM_005245828.3 | XP_005245885.1 | 5 | 1316 | |||
XM_005245830.3 | XP_005245887.1 | 5 | 1197 | |||
XM_005245832.3 | XP_005245889.1 | 5 | 1222 | |||
XM_006710555.3 | XP_006710618.1 | 6 | 1400 | |||
XM_011541198.2 | XP_011539500.1 | 5 | 1219 | |||
XM_017000954.1 | XP_016856443.1 | 5 | 1319 | |||
XM_017000955.1 | XP_016856444.1 | 5 | 1197 | |||
XM_017000956.1 | XP_016856445.1 | 5 | 1194 | |||
XM_017000957.1 | XP_016856446.1 | 5 | 1319 | |||
XM_017000958.1 | XP_016856447.1 | 5 | 1299 | |||
XM_017000959.1 | XP_016856448.1 | 5 | 1299 | |||
GenBank mRNA | BC028731.2 | - | 5 | 1143 | 4829037 | |
BC029505.1 | - | 5 | 1180 | 5273528 |