Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257378.1 | NP_001244307.1 | 8 | 813 | ||
NM_001257379.1 | NP_001244308.1 | 7 | 643 | |||
NM_001257382.1 | NP_001244311.1 | 7 | 713 | |||
NM_174975.4 | NP_777635.1 | 6 | 595 | |||
XM_011530128.2 | XP_011528430.1 | 6 | 563 | |||
GenBank mRNA | AK131358.1 | - | 7 | 617 | ||
AY158086.1 | - | 6 | 506 | |||
AY240872.1 | - | 6 | 595 | |||
BC069641.1 | - | 6 | 518 | 7262444 | ||
BC101001.2 | - | 8 | 752 | 40006372 | ||
BC101002.1 | - | 7 | 582 | 40006375 | ||
BC101003.1 | - | 7 | 652 | 40006376 | ||
BC101004.1 | - | 6 | 534 | 40006377 |