Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG2 SIRNA Assays ›
Gene Symbol:
DLG2
Gene Name:
discs large MAGUK scaffold protein 2
Gene Aliases:
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location:
Chr.11: 83455009 - 85628534 on Build GRCh38
UniGene ID:
Hs.367656
Species:
Human
siRNA ID
HSS138272
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142699.1
NP_001136171.1
28
6370
NM_001142700.1
NP_001136172.1
20
5405
NM_001142702.1
NP_001136174.1
11
4447
NM_001206769.1
NP_001193698.1
22
6054
NM_001300983.1
NP_001287912.1
23
5992
NM_001364.3
NP_001355.2
23
6046
XM_005273810.4
XP_005273867.1
22
5689
XM_005273811.4
XP_005273868.1
22
5635
XM_011544778.2
XP_011543080.1
29
7661
XM_011544780.2
XP_011543082.1
29
7607
XM_011544782.2
XP_011543084.1
24
6132
XM_011544788.2
XP_011543090.1
21
5479
XM_017017254.1
XP_016872743.1
30
7703
XM_017017255.1
XP_016872744.1
29
7664
XM_017017256.1
XP_016872745.1
29
7193
XM_017017257.1
XP_016872746.1
28
8125
XM_017017258.1
XP_016872747.1
27
7301
XM_017017259.1
XP_016872748.1
27
7022
XM_017017260.1
XP_016872749.1
26
7954
XM_017017261.1
XP_016872750.1
28
7451
XM_017017262.1
XP_016872751.1
23
6156
XM_017017263.1
XP_016872752.1
25
6174
XM_017017264.1
XP_016872753.1
24
6351
XM_017017265.1
XP_016872754.1
24
6348
XM_017017266.1
XP_016872755.1
22
6058
XM_017017267.1
XP_016872756.1
24
6256
XM_017017268.1
XP_016872757.1
24
6077
XM_017017269.1
XP_016872758.1
24
6778
XM_017017270.1
XP_016872759.1
24
6078
XM_017017271.1
XP_016872760.1
23
6255
XM_017017272.1
XP_016872761.1
23
6252
XM_017017273.1
XP_016872762.1
22
6007
XM_017017274.1
XP_016872763.1
24
6202
XM_017017275.1
XP_016872764.1
21
5957
XM_017017276.1
XP_016872765.1
23
6680
XM_017017277.1
XP_016872766.1
23
6000
XM_017017278.1
XP_016872767.1
21
5904
XM_017017279.1
XP_016872768.1
23
5922
XM_017017280.1
XP_016872769.1
23
6599
XM_017017281.1
XP_016872770.1
22
6099
XM_017017283.1
XP_016872772.1
22
5904
XM_017017284.1
XP_016872773.1
22
5576
XM_017017285.1
XP_016872774.1
22
6526
XM_017017286.1
XP_016872775.1
20
5440
XM_017017287.1
XP_016872776.1
13
4460
XM_017017288.1
XP_016872777.1
12
4299
XM_017017289.1
XP_016872778.1
12
5874
XM_017017290.1
XP_016872779.1
12
4519
XM_017017291.1
XP_016872780.1
11
4266
XM_017017292.1
XP_016872781.1
11
5778
GenBank mRNA
AK094585.1
-
1
468
AK125151.1
-
1
1476
AK125152.1
-
1
999
BC104969.1
-
1
567
BX648386.1
-
1
1873
CR933674.1
-
11
4416
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
sensory perception of pain
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
GMP metabolic process
GDP metabolic process
receptor localization to synapse
guanylate kinase activity
protein binding
kinase binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0