Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198827.3 | NP_942122.2 | 11 | 1737 | ||
XM_005253566.2 | XP_005253623.1 | 10 | 1557 | |||
XM_011538203.2 | XP_011536505.1 | 12 | 1833 | |||
XM_011538204.2 | XP_011536506.1 | 12 | 1833 | |||
XM_011538205.2 | XP_011536507.1 | 11 | 1653 | |||
XM_011538206.2 | XP_011536508.1 | 12 | 1833 | |||
XM_011538207.2 | XP_011536509.1 | 12 | 1833 | |||
XM_011538208.2 | XP_011536510.1 | 10 | 1398 | |||
XM_011538209.2 | XP_011536511.1 | 12 | 1833 | |||
XM_011538210.2 | XP_011536512.1 | 12 | 1833 | |||
XM_011538211.2 | XP_011536513.1 | 4 | 323 | |||
XM_011538212.2 | XP_011536514.1 | 2 | 118 | |||
GenBank mRNA | AK131538.1 | - | 3 | 491 | ||
AY255587.1 | - | 2 | 116 | |||
AY532280.1 | - | 11 | 1351 | |||
BC112307.1 | - | 11 | 1208 | 8327775 | ||
BC112309.1 | - | 11 | 1208 | 8327777 | ||
BC143775.1 | - | 12 | 1304 | 9052289 | ||
EU523118.1 | - | 11 | 1252 |
Set Membership: |
![]() |