Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CES4A SIRNA Assays ›
Gene Symbol:
CES4A
Gene Name:
carboxylesterase 4A
Gene Aliases:
CES6, CES8
Chromosome Location:
Chr.16: 66988589 - 67009758 on Build GRCh38
UniGene ID:
Hs.346947
Species:
Human
siRNA ID
HSS138425
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CES4A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190201.1
NP_001177130.1
3, 4
546
NM_001190202.1
NP_001177131.1
3, 4
558
NM_001318506.1
NP_001305435.1
3, 4
602
NM_173815.6
NP_776176.5
5, 6
865
XM_005255896.4
XP_005255953.1
5, 6
862
XM_011523021.2
XP_011521323.1
6, 7
994
XM_011523023.2
XP_011521325.1
6, 7
982
XM_011523024.2
XP_011521326.1
6, 7
994
XM_011523025.2
XP_011521327.1
6, 7
994
XM_011523026.2
XP_011521328.1
5, 6
874
XM_011523027.2
XP_011521329.1
6, 7
994
XM_011523028.2
XP_011521330.1
5, 6
882
XM_011523029.2
XP_011521331.1
6, 7
994
XM_011523030.2
XP_011521332.1
5, 6
862
XM_017023160.1
XP_016878649.1
6, 7
982
XM_017023161.1
XP_016878650.1
6, 7
982
XM_017023162.1
XP_016878651.1
6, 7
994
XM_017023163.1
XP_016878652.1
6, 7
994
XM_017023164.1
XP_016878653.1
6, 7
982
XM_017023165.1
XP_016878654.1
6, 7
982
XM_017023166.1
XP_016878655.1
6, 7
994
XM_017023167.1
XP_016878656.1
3, 4
946
XM_017023168.1
XP_016878657.1
3, 4
952
GenBank mRNA
AK094783.1
-
3, 4
563
AK293061.1
-
3, 4
394
AK295483.1
-
3, 4
498
AK296064.1
-
5, 6
785
AK300792.1
-
3, 4
506
BC064573.1
-
3, 4
326
BC084555.1
-
4, 5
570
DC306014.1
-
3, 4
546
Back To Top
More Information
Panther Classification:
Molecular Function -
esterase
Gene Ontology Categories:
Process(es)
carboxylic ester hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline