Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143838.2 | NP_001137310.1 | 10 | 1502 | ||
NM_001284509.1 | NP_001271438.1 | 10 | 1451 | |||
NM_001284510.1 | NP_001271439.1 | 9 | 1373 | |||
NM_177550.4 | NP_808218.1 | 10 | 1502 | |||
XM_011523795.2 | XP_011522097.1 | 10 | 1492 | |||
GenBank mRNA | AK127797.1 | - | 10 | 1399 | ||
AK172779.1 | - | 3 | 355 | |||
AK172785.1 | - | 10 | 1447 | |||
AY151833.1 | - | 10 | 1426 | |||
BC104795.1 | - | 10 | 1458 | 8143798 | ||
BC112151.1 | - | 10 | 1458 | 8327619 | ||
BC143689.1 | - | 10 | 1458 | 9052203 | ||
CR749584.1 | - | 5 | 616 |