Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031749.2 | NP_001026919.2 | 5 | 720 | ||
NM_001288763.1 | NP_001275692.1 | 4 | 770 | |||
NM_182573.2 | NP_872379.2 | 5 | 692 | |||
XM_011526794.2 | XP_011525096.1 | 5 | 776 | |||
GenBank mRNA | AK055031.1 | - | 5 | 670 | ||
AK301693.1 | - | 5 | 720 | |||
AY358719.1 | - | 5 | 693 | |||
BC057816.1 | - | 5 | 706 | 30345834 | ||
CK823626.1 | - | 4 | 520 |