Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321298.1 | NP_001308227.1 | 8 | 1264 | ||
NM_173637.3 | NP_775908.2 | 7 | 1166 | |||
NR_135612.1 | 7 | 1166 | ||||
XM_011527926.1 | XP_011526228.1 | 7 | 960 | |||
GenBank mRNA | AK097761.1 | - | 8 | 1248 | ||
BC031671.1 | - | 7 | 1166 | 5164630 |