Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FFAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195755.1 | NP_001182684.1 | 3 | 992 | ||
NM_181745.3 | NP_859529.2 | 4 | 1040 | |||
GenBank mRNA | AB115768.1 | - | 4 | 984 | ||
AK301525.1 | - | 3 | 965 | |||
AY288417.1 | - | 4 | 981 | |||
BC101175.2 | - | 3 | 936 | 40016354 |
Set Membership: |
G-protein coupled receptor |